234605
2-Naphthyl acetate
98%
Synonym(s):
β-Naphthyl acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CO2C10H7
CAS Number:
Molecular Weight:
186.21
Beilstein:
1868291
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
Assay
98%
extent of labeling
<0.2% free naphthol
mp
67-70 °C (lit.)
SMILES string
CC(=O)Oc1ccc2ccccc2c1
InChI
1S/C12H10O2/c1-9(13)14-12-7-6-10-4-2-3-5-11(10)8-12/h2-8H,1H3
InChI key
RJNPPEUAJCEUPV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service