Skip to Content
Merck
All Photos(1)

Key Documents

214531

Sigma-Aldrich

Acid Orange 8

Dye content 65 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N2NaO4S
CAS Number:
Molecular Weight:
364.35
Colour Index Number:
15575
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 65%

solubility

water: 1 mg/mL, clear, red-orange

λmax

490 nm

ε (extinction coefficient)

≥10000 at 261-267 nm in water at 0.01499 g/L
≥15000 at 486-492 nm in water at 0.01499 g/L
≥23000 at 226-232 nm in water at 0.01499 g/L
≥6000 at 306-312 nm in water at 0.01499 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1cc(ccc1\N=N\c2c(O)ccc3ccccc23)S([O-])(=O)=O

InChI

1S/C17H14N2O4S.Na/c1-11-10-13(24(21,22)23)7-8-15(11)18-19-17-14-5-3-2-4-12(14)6-9-16(17)20;/h2-10,20H,1H3,(H,21,22,23);/q;+1/p-1/b19-18+;

InChI key

WPWNIQBSYQVEKJ-LTRPLHCISA-M

General description

Acid Orange 8 is an azo dye derivative, that is usually used in textile dyeing.[1]

Application

Acid Orange 8 has been used to study its photodegradation induced by TiO2 (titanium dioxide) in aqueous solution.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titanium dioxide mediated photocatalyzed degradation of a textile dye derivative, acid orange 8, in aqueous suspensions
Saquib M and Muneer M
Desalination, 155(3), 255-263 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service