Skip to Content
Merck
All Photos(1)

Documents

201391

Sigma-Aldrich

Brilliant Blue G

Dye content 50 %

Synonym(s):

Acid blue 90, Coomassie brilliant blue G 250

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H52N3NaO7S2
CAS Number:
Molecular Weight:
858.05
Colour Index Number:
42655
Beilstein:
5230822
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 50%

λmax

610 nm

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3C)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=CC(\C=C5C)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

Looking for similar products? Visit Product Comparison Guide

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service