Skip to Content
Merck
All Photos(1)

Key Documents

195928

Sigma-Aldrich

3-(4-tert-Butylphenoxy)benzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4OC6H4CHO
CAS Number:
Molecular Weight:
254.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.57 (lit.)

bp

152 °C/0.4 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(Oc2cccc(C=O)c2)cc1

InChI

1S/C17H18O2/c1-17(2,3)14-7-9-15(10-8-14)19-16-6-4-5-13(11-16)12-18/h4-12H,1-3H3

InChI key

YZIHNHLGGMSWAD-UHFFFAOYSA-N

Application

3-(4-tert-Butylphenoxy)benzaldehyde was used in high-throughput parallel synthesis of library compounds for early drug discovery[1]. It was also used in purification platform developed by ArQule that is based on reverse phase high performance liquid chromatography separation and mass directed fractionation[2].

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James N Kyranos et al.
Journal of combinatorial chemistry, 6(5), 796-804 (2004-09-14)
High-throughput parallel synthesis of library compounds for early drug discovery requires high-throughput analytical methods to confirm synthesis, identify reaction products, and determine purity. An ultrafast 1.0-min HPLC/UV/ELSD/MS method was developed and compared to our standard 2.5- and 5.0-min methods in
High throughput HPLC/MS purification in support of drug discovery
Goetzinger W, et al.
International Journal of Mass Spectrometry, 238(2), 153-162 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service