Skip to Content
Merck
All Photos(2)

Key Documents

195308

Sigma-Aldrich

3-(4-Chlorophenoxy)benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OC6H4CHO
CAS Number:
Molecular Weight:
232.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

refractive index

n20/D 1.608 (lit.)

bp

125 °C/0.1 mmHg (lit.)

density

1.213 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Oc2cccc(C=O)c2)cc1

InChI

1S/C13H9ClO2/c14-11-4-6-12(7-5-11)16-13-3-1-2-10(8-13)9-15/h1-9H

InChI key

MYQFRCYBOOWGJQ-UHFFFAOYSA-N

General description

3-(4-Chlorophenoxy)benzaldehyde has been investigated for anti-tumour activity[1].

Application

3-(4-Chlorophenoxy)benzaldehyde was used in the synthesis of fatty acid amide hydrolase inhibitors bearing spirocyclic diamine cores[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Bogatcheva et al.
Bioorganic & medicinal chemistry letters, 20(1), 201-205 (2009-11-18)
As part of our ongoing research effort to develop new therapeutics for treatment of tuberculosis (TB), we synthesized a combinatorial library of 10,358 compounds on solid support using a pool-and-split technique and tested the resulting compounds for activity against Mycobacteriumtuberculosis.
Heteroarylureas with Spirocyclic Diamine Cores as Inhibitors of Fatty Acid Amide Hydrolase.
Keith JM, et al.
Bioorganic & Medicinal Chemistry Letters (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service