Skip to Content
Merck
All Photos(1)

Key Documents

193518

Sigma-Aldrich

3-Chlorophenethyl alcohol

98%

Synonym(s):

2-(3-Chlorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CH2OH
CAS Number:
Molecular Weight:
156.61
Beilstein:
2042059
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.549 (lit.)

bp

135-137 °C/13 mmHg (lit.)

density

1.181 g/mL at 25 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCCc1cccc(Cl)c1

InChI

1S/C8H9ClO/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6,10H,4-5H2

InChI key

NDWAVJKRSASRPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chlorophenethyl alcohol undergoes coupling reaction with dibutylamine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark R Biscoe et al.
Journal of the American Chemical Society, 130(21), 6686-6687 (2008-05-02)
A new class of one-component Pd precatalysts bearing biarylphosphine ligands is described. These precatalysts are air- and thermally stable, are easily activated under normal reaction conditions at or below room temperature, and ensure the formation of the highly active monoligated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service