Skip to Content
Merck
All Photos(1)

Key Documents

180777

Sigma-Aldrich

N,N-Dimethyl-m-toluidine

97%

Synonym(s):

3-(Dimethylamino)toluene, 3-Methyl-N,N-dimethylaniline, Dimethyl-m-toluidine, N,N,3-Trimethylaniline, N,N,3-Trimethylbenzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N(CH3)2
CAS Number:
Molecular Weight:
135.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.55 (lit.)

bp

215 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)c1cccc(C)c1

InChI

1S/C9H13N/c1-8-5-4-6-9(7-8)10(2)3/h4-7H,1-3H3

InChI key

CWOMTHDOJCARBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethyl-m-toluidine may be used in chemical synthesis studies.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service