17781
α,α′-Diglycerol
technical, ≥85%
Synonym(s):
3,3′-Oxydi(1,2-propanediol), Bis(2,3-dihydroxypropyl) ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
O(CH2CHOHCH2OH)2
CAS Number:
Molecular Weight:
166.17
Beilstein:
1904420
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
grade
technical
Assay
≥85%
refractive index
n20/D 1.489
density
1.280 g/mL at 20 °C (lit.)
SMILES string
OCC(O)COCC(O)CO
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service