Skip to Content
Merck
All Photos(1)

Documents

175080

Sigma-Aldrich

Magic Acid

100%

Synonym(s):

Fluorosulfuric acid-antimony pentafluoride 1:1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FSO3H·SbF5
CAS Number:
Molecular Weight:
316.82
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Assay

100%

storage temp.

−20°C

SMILES string

OS(F)(=O)=O.F[Sb](F)(F)(F)F

InChI

1S/FHO3S.5FH.Sb/c1-5(2,3)4;;;;;;/h(H,2,3,4);5*1H;/q;;;;;;+5/p-5

InChI key

QNDPUZFBWUBSNH-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Pereira Marques et al.
Clinical oral investigations, 22(9), 2989-2996 (2018-02-18)
The aim of this study was to evaluate the clinical performance of silorane-based composite restorations applied after different surface treatments. This controlled and randomized clinical trial included 26 patients with class I restoration indications. The teeth were randomly assigned to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service