Skip to Content
Merck
All Photos(1)

Documents

163341

Sigma-Aldrich

2-Nitrophenylacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2CN
CAS Number:
Molecular Weight:
162.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

82-85 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1CC#N

InChI

1S/C8H6N2O2/c9-6-5-7-3-1-2-4-8(7)10(11)12/h1-4H,5H2

InChI key

YPRFCQAWSNWRLM-UHFFFAOYSA-N

Application

2-Nitrophenylacetonitrile was used in synthesis of 2 or 2,4-substituted α-carbolines via a one-pot tandem reaction of α,β-unsaturated ketones. It was used in the synthesis of strychnine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofei Zhang et al.
Organic & biomolecular chemistry, 12(2), 355-361 (2013-11-23)
An effective and convenient method has been developed for the preparation of 2 or 2,4-substituted α-carbolines via a one-pot tandem reaction of α,β-unsaturated ketones with 2-nitrophenylacetonitrile in the presence of zinc dust, acetic acid and triethylamine. This protocol presents a
Takashi Ohshima et al.
Journal of the American Chemical Society, 124(49), 14546-14547 (2002-12-06)
The enantioselective total synthesis of (-)-strychnine was accomplished through the use of the highly practical catalytic asymmetric Michael reaction (0.1 mol % of (R)-ALB, more than kilogram scale, without chromatography, 91% yield and >99% ee) as well as a tandem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service