Skip to Content
Merck
All Photos(1)

Key Documents

156574

Sigma-Aldrich

1,3-Dibromobutane

97%

Synonym(s):

1,3-Butylenebromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHBrCH2CH2Br
CAS Number:
Molecular Weight:
215.91
Beilstein:
1731231
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.5080 (lit.)

bp

175 °C (lit.)

density

1.8 g/mL at 25 °C (lit.)

SMILES string

CC(Br)CCBr

InChI

1S/C4H8Br2/c1-4(6)2-3-5/h4H,2-3H2,1H3

InChI key

XZNGUVQDFJHPLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

It has been to investigate its molecular structure and conformational composition by gas-phase electron diffraction and ab initio methods.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Structure and Conformational Composition of 1,3-Dichlorobutane and 1,3-Dibromobutane. A Gas-Phase Electron Diffraction and ab Initio Investigation.
Aarset K, et al.
The Journal of Physical Chemistry, 99(28), 11089-11094 (1995)
Rahul A Patil et al.
Analytical and bioanalytical chemistry, 410(19), 4645-4655 (2018-01-31)
The rapid expansion in the study and use of ionic liquids (ILs) is a result of their unique properties including negligible volatility, high thermal stability, and ability to dissolve disparate compounds. However, because ILs have infinitely variable structures (often referred

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service