Skip to Content
Merck
All Photos(1)

Key Documents

15288

Sigma-Aldrich

Dimethyl (Boc-amino)malonate

≥95% (HPLC)

Synonym(s):

(Boc-amino)malonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHCH(COOCH3)2
CAS Number:
Molecular Weight:
247.25
Beilstein:
2380322
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

refractive index

n20/D 1.444

density

1.148 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COC(=O)C(NC(=O)OC(C)(C)C)C(=O)OC

InChI

1S/C10H17NO6/c1-10(2,3)17-9(14)11-6(7(12)15-4)8(13)16-5/h6H,1-5H3,(H,11,14)

InChI key

UIQVFHUDWIETEY-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service