Skip to Content
Merck
All Photos(2)

Key Documents

145998

Sigma-Aldrich

2-Quinoxaloyl chloride

Synonym(s):

2-Quinoxalinecarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5ClN2O
CAS Number:
Molecular Weight:
192.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

113-115 °C (lit.)

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

ClC(=O)c1cnc2ccccc2n1

InChI

1S/C9H5ClN2O/c10-9(13)8-5-11-6-3-1-2-4-7(6)12-8/h1-5H

InChI key

SOPDQKNXOCUBSR-UHFFFAOYSA-N

General description

2-Quinoxaloyl chloride reacts with chiral α-hydroxy carboxylic acids to yield UV and fluorescent derivatives.

Application

2-Quinoxaloyl chloride was used in the synthesis of novel DNA interactive quinoxaline-carbohydrate hybrids possessing disaccharides as the carbohydrate moieties.
Reactant involved in the synthesis of a variety of inhibitors including:
  • Gelatinase inhibitors for cancer treatments
  • mGluR5 non-competitve antagonists
  • Heterocyclic analogs used as SIRT1 activators
  • 3rd Generation multidrug resistance modulators

Reactant involved in preparation of PET ligants for breast cancer resistance protein imaging

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC resolution of hydroxyl carboxylic acid enantiomers using 2-quinoxaloyl chloride as a new precolumn derivatizing agent.
Brightwell M, et al.
Journal of Liquid Chromatography and Related Technologies, 18(14), 2765-2781 (1995)
Kazunobu Toshima et al.
Bioorganic & medicinal chemistry letters, 14(11), 2777-2779 (2004-05-06)
The novel DNA interactive quinoxaline-carbohydrate hybrids possessing disaccharides as the carbohydrate moieties were designed and synthesized, and their DNA photocleaving abilities were evaluated in order to examine the effect of the disaccharide structures. The configurations of the glycosidic bonds in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service