145815
Adenine sulfate
97%
Synonym(s):
Adenine hemisulfate salt, 6-Aminopurine hemisulfate salt, Adenine sulfate salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C5H5N5 · 1/2H2SO4
CAS Number:
Molecular Weight:
184.17
Beilstein:
3820263
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
Assay
97%
SMILES string
OS(O)(=O)=O.Nc1ncnc2[nH]cnc12.Nc3ncnc4[nH]cnc34
InChI
1S/2C5H5N5.H2O4S/c2*6-4-3-5(9-1-7-3)10-2-8-4;1-5(2,3)4/h2*1-2H,(H3,6,7,8,9,10);(H2,1,2,3,4)
InChI key
LQXHSCOPYJCOMD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service