Skip to Content
Merck
All Photos(1)

Key Documents

116815

Sigma-Aldrich

Acetoxytrimethyllead(IV)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2Pb(CH3)3
CAS Number:
Molecular Weight:
311.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

mp

180 °C (dec.) (lit.)

SMILES string

CC(=O)O[Pb](C)(C)C

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Holden et al.
Archives of biochemistry and biophysics, 291(1), 187-194 (1991-11-15)
The three-dimensional conformation of a protein provides a wealth of biochemical information and with the advent of cloning techniques that allow the preparation of proteins almost at will, a renewed interest has arisen in the crystallographic determination of protein structures.
P D Sun et al.
Acta crystallographica. Section D, Biological crystallography, 56(Pt 2), 161-168 (2000-02-10)
A heavy-atom screening method aided by mass spectrometry is described here. Using mass spectrometry, several heavy-atom compounds have been screened in order to obtain potential phasing derivatives for the crystals of a human immunoglobulin Fc receptor, Fc gamma RIII. Of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service