Skip to Content
Merck
All Photos(1)

Key Documents

AABH93DE34A0

AA BLOCKS, INC.

Lithium bis(1,1,2,2,3,3,4,4,4-nonafluoro-1-butanesulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
587.14
MDL number:

Iupac Name

lithium bis((perfluorobutyl)sulfonyl)amide

Inchi Code

1S/C8F18NO4S2.Li/c9-1(10,5(17,18)19)3(13,14)7(23,24)32(28,29)27-33(30,31)8(25,26)4(15,16)2(11,12)6(20,21)22;/q-1;+1

InChI key

QIWLMMWTZVIAFK-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

95.0%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/119229-99-1

Synonym

Lithium Bis(nonafluorobutanesulfonyl)imide

MSDS

https://www.aablocks.com/downloadSds?cas=119229-99-1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library