Merck
  • Zinc Iodide Dimethyl Sulfoxide Reduces Collagen Deposition by Increased Matrix Metalloproteinase-2 Expression and Activity in Lung Fibroblasts.
Zinc Iodide Dimethyl Sulfoxide Reduces Collagen Deposition by Increased Matrix Metalloproteinase-2 Expression and Activity in Lung Fibroblasts.
Biomedicines (2024-06-27)
Michael Roth, Bo Han, Chong Teck S'ng, Ba Xuan Hoang, Christopher Lambers
ABSTRACT
Chronic inflammatory lung diseases are characterized by disease-specific extracellular matrix accumulation resulting from an imbalance of matrix metalloproteinases (MMPs) and their inhibitors. Zinc is essential for the function of MMPs, and zinc deficiency has been associated with enhanced tissue remodeling. This study assessed if zinc iodide (ZnI) supplementation through dimethyl sulfoxide (DMSO) modifies the action of MMPs in isolated human lung fibroblasts. The expression and activity of two gelatinases, MMP-2 and MMP-9, were determined by gelatin zymography and enzyme-linked immuno-sorbent assay (ELISA). Collagen degradation was determined by cell-based ELISAs. Collagen type I and fibronectin deposition was stimulated by human recombinant tumor growth factor β1 (TGF-β1). Untreated fibroblasts secreted MMP-2 but only minute amounts of MMP-9. TGF-β1 (5 ng/mL) reduced MMP-2 secretion, but stimulated collagen type I and fibronectin deposition. All the effects of TGF-β1 were significantly reduced in cells treated with ZnI-DMSO over 24 h, while ZnI and DMSO alone had a lower reducing effect. ZnI-DMSO alone did not increase MMP secretion but enhanced the ratio of active to inactive of MMP-2. ZnI alone had a lower enhancing effect than ZnI-DMSO on MMP activity. Furthermore, MMP-2 activity was increased by ZnI-DMSO and ZnI in the absence of cells. Soluble collagen type I increased in the medium of ZnI-DMSO- and ZnI-treated cells. Blocking MMP activity counteracted all the effects of ZnI-DMSO. Conclusion: The data suggest that the combination of ZnI with DMSO reduces fibrotic processes by increasing the degradation of collagen type I by up-regulating the activity of gelatinases. Thus, the combination of ZnI with DMSO might be considered for treatment of fibrotic disorders of the lung. DMSO supported the beneficial effects of ZnI.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MMP-2/MMP-9 Inhibitor I, The MMP-2/MMP-9 Inhibitor I, also referenced under CAS 193807-58-8, controls the biological activity of MMP-2/MMP-9. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.