Merck
  • Synthesis of new UV-B light absorbents: (Acetylphenyl)glycosides with antioxidant activities.
Synthesis of new UV-B light absorbents: (Acetylphenyl)glycosides with antioxidant activities.
Bioorganic & medicinal chemistry letters (2008-05-31)
Takashi Otani, Tetsu Tsubogo, Naoki Furukawa, Takao Saito, Katsumi Uchida, Kanako Iwama, Yuki Kanai, Hirofumi Yajima
ABSTRACT
m-Acetylphenyl-beta-d-glucopyranosides and m-acetylphenyl-alpha/beta-d-mannopyranosides were synthesized by the Koenigs-Knorr, Mitsunobu, and Helferich reactions as key glycosylation reactions, respectively. Their spectroscopic properties and antioxidative activities were characterized as potential ultraviolet B-ray absorbents.
Sigma-Aldrich
3′-Hydroxyacetophenone, 97%
Sigma-Aldrich
3′-Hydroxyacetophenone, ≥99%