Skip to Content
Merck
All Photos(1)

Key Documents

69131

Supelco

Diethyl sebacate

analytical standard

Synonym(s):

Decanedioic acid 1,10-diethyl ester, NSC 8911, Sebacic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)8COOC2H5
CAS Number:
Molecular Weight:
258.35
Beilstein:
1790779
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.436 (lit.)

bp

312 °C (lit.)

mp

1-2 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(=O)CCCCCCCCC(=O)OCC

InChI

1S/C14H26O4/c1-3-17-13(15)11-9-7-5-6-8-10-12-14(16)18-4-2/h3-12H2,1-2H3

InChI key

ONKUXPIBXRRIDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service