Skip to Content
Merck
All Photos(1)

Key Documents

13369-U

Supelco

Carbon Adsorbent Sampler Kit

matrix CMS/SGPC Kit II

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23201100
NACRES:
SB.54

product line

Carboxen®
Graphsphere

form

powder

packaging

kit of 9 bottles

technique(s)

LPLC: suitable

matrix

CMS/SGPC Kit II
Carbon Molecular Sieve (CMS) and Spherical Graphitized Polymer Carbon (SGPC) adsorbents

matrix active group

carbon

particle size

20-45 mesh

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Choosing the right adsorbent or combination of adsorbents can be difficult:
  • Must choose one or more to retain specific analyte(s)
  • May be equally important to release analyte(s) during desorption

This convenient kit allows a cost-effective way to evaluate several of our specialty carbon adsorbents. Once you find an adsorbent or adsorbents that serve your purpose, we will work with you to either make your device for you, or to supply you bulk adsorbent (in a bottle, jar, or ampule; not packed inside a column or device).

For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon

Other Notes

Purchase of this kit is limited to two per customer per calendar year.

Footnote

Each bottle contains 5 g of a different material.

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
Graphsphere is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • 20/45 Carboxen® 563 5 g

  • 20/45 Carboxen 564 5 g

  • 20/45 Carboxen 569 5 g

  • 20/45 Carboxen 572 5 g

  • 20/45 Carboxen 1005 5 g

  • 20/45 Carboxen 1032 5 g

  • 20/45 Carboxen 1033 5 g

  • 20/45 Carboxen 1034 5 g

  • 20/45 Graphsphere 2017 5 g

See All (9)

Storage Class Code

13 - Non Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Synthetic CMS carbons offer tailored adsorbents for specific applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service