Key Documents
TMO4115-2800
Nalgene® sterile, single-use Erlenmeyer flask with vented closure
PETG, bottle volume 2800 mL, case of 4 ea
Synonym(s):
Nalgene Erlenmeyer flask, single use plastic flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.13
material
PETG
sterility
sterile
feature
vented
packaging
case of 4 ea
manufacturer/tradename
Nalgene 4115-2800
bottle volume
2800 mL
General description
Save preparation time and avoid contamination risk by using the Nalgene® Single-Use PETG Erlenmeyer Flasks, which are available with a flat or baffled bottom. These flasks are ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available with a vented or non-vented closure.
Features and Benefits
  • Sterile and single us, reducing contamination risk
  • BPA-free
  • Collapses when autoclaved, reducing bio hazardous waste
  • Graduated for quick volume assessment
  • Increases user safety due to the break-resistant and robust design of the crystal-clear PETG material
  • Facilitates visual inspection and monitoring of contents due to its crystal-clear composition
  • Equipped with a vented cap to support gas exchange during processes like cell culture
Other Notes
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)