Skip to Content
Merck
All Photos(1)

Key Documents

SRP8034

Sigma-Aldrich

IL-2 (human): FC (human)

recombinant, expressed in HEK 293 cells, >95% (SDS-PAGE)

Synonym(s):

Aldesleukin, INN, Interleukin-2, T-cell growth factor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

human

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~42 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.01 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL2(3558)

General description

Interleukin-2 (IL-2) is a 133 amino acid glycoprotein with one intramolecular disulfide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils and induces mononuclear cells to secrete IFN-γ and TNF-α and -β. Moreover, studies have shown that IL-2 is required for activation-induced apoptosis, an important homeostatic mechanism in the immune system, which is involved in the maintenance of peripheral tolerance to self-antigens.IL-2 promotes T cell proliferation and particularly naive T cells. IL-2 signaling on activated T cells is effected through a quaternary high-affinity receptor complex consisting of IL-2, IL-2Rα (CD25), IL-2Rβ and IL-2Rγ. Naive T cells are relatively insensitive to IL-2 as they only express small amounts of IL-2Rβ and IL-2Rγ. They only acquire sensitivity after CD25 expression, which captures the cytokine and presents it to the IL-2Rβ and IL-2Rγ receptors. IL-2 Superkine (Fc) is an artificial variant of IL-2 containing mutations at positions L80F / R81D / L85V / I 86V / I92F. These mutations are located in the molecule′s core that acts to stabilize the structure and to give it a receptor-binding conformation mimicking native IL-2 bound to CD25. These mutations effectively eliminate the functional requirement of IL-2 for CD25 expression and elicit proliferation of T cells. Compared to IL-2, the IL-2 superkine induces superior expansion of cytotoxic T cells, leading to improved antitumour responses in vivo, and elicits proportionally less toxicity by lowering the expansion of Tregulatory cells and reducing pulmonary oedema.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

The extracellular domain of human IL-2 (aa 21-153) (mutant H9 containing the mutations L80F / R81D / L85V / I 86V / I92F) is fused at the C-terminus to the Fc portion of human IgG2.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service