Skip to Content
Merck
All Photos(1)

Key Documents

SRP3153

Sigma-Aldrich

SCGF-beta human

≥98% (SDS-PAGE & HPLC), recombinant, expressed in E. coli, lyophilized, suitable for cell culture

Synonym(s):

Stem Cell Growth Factor-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Pricing and availability is not currently available.

product name

SCGF-beta human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤7.0 ng/mL ED50

mol wt

25.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

General description

SCGF alpha and beta are hematopoietic growth factor that exerts their activity at early stages of hematopoiesis. The SCGFs are non-glycosylated species-specific cytokines that can support growth of primitive hematopoietic cells, and in combination with EPO or GM-CSF, promote proliferation or erthroid or myeloid progenitors, respectively. Recombinant human SCGF-b is a 25.0 kDa polypeptide containing 227 amino acid residues.

Biochem/physiol Actions

SCGF alpha and beta are hematopoietic growth factor that exerts their activity at early stages of hematopoiesis. Recombinant human SCGF-b is a 25.0 kDa polypeptide containing 227 amino acid residues.

Sequence

MGHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ PSASPHPLSP DQPNGGTLEN CVAQASDDGS WWDHDCQRRL YYVCEFPF

Physical form

Lyophilized from 0.25x PBS + 1mM DTT.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Ding et al.
Fish & shellfish immunology, 33(3), 494-503 (2012-06-12)
The toll-like receptors (TLRs) are an important gene family in host innate immunologic surveillance. The TLR22 gene is an essential member of the TLRs that is only found in aquatic animals and has been detected in some bony fish. Here

Articles

Read article on hematopoietic cytokines and hematopoiesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service