Skip to Content
Merck
All Photos(1)

Key Documents

SML2870

Sigma-Aldrich

BI 99179

≥98% (HPLC)

Synonym(s):

(1R,3S)-3-Propionylaminocyclopentanecarboxylic acid N-[4-(benzoxazol-2-yl)phenyl]-N-methylamide, (1R,3S)-N-[4-(2-Benzoxazolyl)phenyl]-N-methyl-3-[(1-oxopropyl)amino]cyclopentanecarboxamide, BI99179

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N3O3
CAS Number:
Molecular Weight:
391.46
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCC(N[C@H]1CC[C@@H](C(N(C)C2=CC=C(C=C2)C3=NC4=C(O3)C=CC=C4)=O)C1)=O

InChI key

YNFDIGJKJPNFFD-SJORKVTESA-N

Biochem/physiol Actions

BI 99179 is a non-cytotoxic (by LDH release; 30 μM, U937) orally available, brain-penetrant, highly potent and selective fatty acid synthase (FAS, FASN) inhibitor (IC50 = 79 nM by cell-free human FAS assay) with little or no inhibitory potency toward a panel of 30 receptors & channels (<20% inhibition at 10 μM) and human CYP450 isoenzymes (1A2/2C9/2C19/2D6/3A4 IC50 >10 μM). BI 99179 inhibits cellular FAS activity in vitro (IC50 = 180 nM/human H1975, 600 nM/murine hypothalamic N-42 cells) and increases hypothalamic [malonyl-CoA] in rats in vivo (2 h post 10 mg/kg or 2-24 hr post 100 mg/kg p.o.). Note: adverse effects in rats are reported at day 4 (30 mg/kg) and day 9 (3 mg/kg).
Non-cytotoxic, orally available, brain-penetrant, highly potent and selective fatty acid synthase (FAS, FASN) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jörg T Kley et al.
Bioorganic & medicinal chemistry letters, 21(19), 5924-5927 (2011-08-30)
Based on a high-throughput screen, cyclopentanecarboxanilides were identified as a new chemotype of non-covalent inhibitors of type I fatty acid synthase (FAS). Starting from initial hits we aimed at generating a tool compound suitable for the in vivo validation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service