Skip to Content
Merck
All Photos(3)

Key Documents

SAE0116

Sigma-Aldrich

Heparanase-1 pre-activated human

recombinant, expressed in HEK 293 cells

Synonym(s):

Endo-glucoronidase, HPA, HPA1, HPR1, HPSE, HPSE1, HSE1, Heparanase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

Assay

≥95.0% (SDS-PAGE)

form

lyophilized powder

impurities

≤1.00 EU/μg (LAL)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

General description

Recombinant human heparanase-1 is expressed in human HEK 293 cells as an active heterodimer glycoprotein containing a 50 kDa subunit and a 8 kDa subunit. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher specific activity of the protein. The protein is produced with no artificial tags.

Biochem/physiol Actions

Heparanase-1 (heparanase) is the only heparan sulfate degrading endoglycosidase. It is involved in the regulation of multiple biological processes that increase tumor growth, metastasis, angiogenesis and inflammation. Heparanase is involved in ECM remodeling and heparan sulfate-linked biological molecules release, including cytokines and growth factors. Heparanase cleaves the heparan sulfate side chains of heparan sulfate proteoglycans into fragments of 10–20 sugar units.
Heparanase is initially translated as a preproenzyme containing a signal peptide. Cleavage of the signal sequence yields a latent 65 kDa pro-heparanase, which must undergo further processing for activity. Mature active heparanase is a heterodimer consisting an N-terminal 8 kDa subunit and a C-terminal 50 kDa subunit.

Unit Definition

One heparanase activity unit is the amount of heparanase required to cleave 1 pmol of pentasaccharide to disaccharide and trisaccharide in one minute, at 37 °C, pH 5.0.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service