Documents
EP0030128974
Eppendorf® Twin tec PCR plates, 384 wells
red polypropylene plate, conical bottom colorless wells, pkg of 300 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material
conical bottom colorless wells
red polypropylene plate
sterility
non-sterile
feature
skirt (stackable)
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128974
technique(s)
PCR: suitable
maximum volume
-45 μL
size
384 wells
well maximum volume
40 μm
binding type
low binding surface
General description
The “Eppendorf” Twin tec PCR plates are designed for single-use PCR applications. The plates feature thin-walled polypropylene wells, designed for optimum heat transfer to samples. Additionally, the plates incorporate sturdy polycarbonate frames to ensure durability, rigidity, automation, and simplified handling in manual workflows. The raised well rims facilitate effective sealing as well as minimize the risk of cross-contamination. These plates are free of common PCR contaminants such as DNA, DNAse, RNAse, and PCR inhibitors.
Application
“Eppendorf” Twin tec PCR plates, 384 wells are suitable for:
  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling
Features and Benefits
  • “Eppendorf” Twin tec PCR plates have an optimized rim suitable for barcodes and automated work processes.
  • Stackable and designed to fit within SBS footprint recommendations.
  • Exceptional solidity and resistance to torque with its polycarbonate frame.
  • These plates are well-suited for most common 384-well thermocyclers.
  • Has Improved well-to-well tolerance.; Each well has a maximum volume of 40 µL.
  • Barcode options are available for skirted and semi-skirted plate types.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG