CLS480124
Corning® LSE™ single block, for Corning LSE single block digital dry baths
single block, 12 x 15 mL centrifuge tubes, 1/cs
Synonym(s):
equipment replacement accessory
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NB.15
Recommended Products
feature
Holds 12 x 15 mm tubes
packaging
pack of 1
case of 2
manufacturer/tradename
Corning 480124
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
Related Categories
General description
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
CLS6785SB, CLS6786SB, CLS6787SB
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)