Skip to Content
Merck
All Photos(1)

Key Documents

HKT-1097

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

hamster kidney

packaging

tube of 5 μg 98061003-DNA-5UG
pkg of vial of cells 98061003-1VL

growth mode

Adherent

karyotype

Modal no. 44

morphology

Fibroblast-like

products

Not specified

receptors

EGF

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Syrian Hamster kidney

Cell Line Description

The cell line HKT-1097 has been established from a 14 month old male Syrian Hamster (Mesocricetus auratus) kidney tumour which has been induced by diethylstilbestrol exposure. HKT-1097 cells express epidermal growth factor (EGF) receptor, Erb-B2 and Erb-B3 as well as vimentin, desmin, glial fibrillary acidic protein (GFAP), TGF alpha, PGP 9.5 and S100 protein and neuron-specific enolase. Karyotype analysis revealed a modal number of 44 chromosomes (41% of cells) and polyploidy in approximately 2% of cells. The cells have a doubling time of about 36 hours.

DNA Profile

Not specified

Culture Medium

DMEM + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4, i.e., seeding at 1 x 10,000 cells / cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Form a multipolar, loosely packed monolayer.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service