Skip to Content
Merck
All Photos(1)

Documents

Ri-1

96090512, human blood, Lymphoblastoid

Synonym(s):

Ri-1 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Ri-1, 96090512

biological source

human blood

growth mode

Suspension

karyotype

92XXXX/69XXX,t(1,2)(p32,q34)t(2,8)(p11,q24)t(14,18

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian B-cell lymphoma

Cell Line Description

The cell line Ri-1 has been established from the peripheral blood of a 48-year-old female patient with B-cell lymphoma. This EBV-negative cell line with 14q+ abnormality was shown to be at an intermediate stage of B-cell differentiation. Ri-1 expresses surface and cytoplasmic immunoglobulin (IgM kappa), HLA-DR, CD20, CD38 and FMC7. Rearranged heavy-light chain genes and one rearranged kappa light-chain gene was detected as well as a kappa and lambda light-chain gene in germ-line configuration.

Application

Study of B-cell lymphoma-leukaemia

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cell growth is very slow after resuscitation; large amounts of debris present.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service