Skip to Content
Merck
All Photos(1)

Documents

RAMOS Cell Line human

85030802, human lymph, Lymphoblast

Synonym(s):

RA 1 Cells, Ramos (RA 1) Cells, Ramos 1 Cells, Ramos-RA1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RAMOS Cell Line human, 85030802

biological source

human lymph

description

Human Caucasian Burkitt′s lymphoma

growth mode

Suspension

karyotype

Modal no. 45

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian Burkitt′s lymphoma

Cell Line Description

Derived from a Burkitt′s lymphoma which does not possess the EBV genome. EBV infectability and permanent conversion into EBV positive sub-lines is possible by in vitro infection. The cells have B lymphocyte characteristics, with surface associated mu and kappa chains.

Application

Tumourigenicity and virus studies: Epstein-Barr virus (EBV).
RAMOS Cell Line human has been used to carry out a MTT (colorimetric assay) cell viability assay to measure calcium ionophore-induced cell death in Ramos cells under different conditions and to determine the expression of ZFP36 ring finger protein like 1 (ZFP36L1) in Ramos B cells during different stages of B cell differentiation.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 13,14
D16S539: 10,13
D5S818: 7,12
D7S820: 11
THO1: 7,9.3
TPOX: 8,9
vWA: 15,16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service