PHR3121
Quetiapine Related Compound B
pharmaceutical secondary standard, certified reference material
Synonym(s):
Norquetiapine dihydrochloride, 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine dihydrochloride
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1592737
API family
quetiapine
CofA
current certificate can be downloaded
packaging
pkg of 40 mg
application(s)
pharmaceutical
storage temp.
-10 to -25°C
SMILES string
S1c2c(cccc2)N=C(c4c1cccc4)N3CCNCC3.Cl.Cl
InChI
1S/C17H17N3S.2ClH/c1-3-7-15-13(5-1)17(20-11-9-18-10-12-20)19-14-6-2-4-8-16(14)21-15;;/h1-8,18H,9-12H2;2*1H
InChI key
PZQCQHZDUIIKFU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service