Skip to Content
Merck
All Photos(1)

Key Documents

PHR1809

Supelco

Tacrolimus

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

FK-506 monohydrate, Tacrolimus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H69NO12 · H2O
CAS Number:
Molecular Weight:
822.03
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1642802

API family

tacrolimus

form

powder

CofA

current certificate can be downloaded

packaging

pkg of 150 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

storage temp.

-10 to -25°C

SMILES string

O.CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)[C@H]2OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@]4(O)O[C@H]([C@H](C[C@@H](C)C\C(C)=C\[C@@H](CC=C)C(=O)C[C@H](O)[C@H]2C)OC)[C@H](C[C@H]4C)OC

InChI

1S/C44H69NO12.H2O/c1-10-13-31-19-25(2)18-26(3)20-37(54-8)40-38(55-9)22-28(5)44(52,57-40)41(49)42(50)45-17-12-11-14-32(45)43(51)56-39(29(6)34(47)24-35(31)48)27(4)21-30-15-16-33(46)36(23-30)53-7;/h10,19,21,26,28-34,36-40,46-47,52H,1,11-18,20,22-24H2,2-9H3;1H2/b25-19+,27-21+;/t26-,28+,29+,30-,31+,32-,33+,34-,36+,37-,38-,39+,40+,44+;/m0./s1

InChI key

NWJQLQGQZSIBAF-MLAUYUEBSA-N

Gene Information

human ... FKBP1A(2280)

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Tacrolimus belongs to the class II drugs of the biopharmaceutics classification system (BCS). Also known as FK-506, it is isolated from Streptomyces tsukubaensis. It is used for its immunosuppressive properties to prevent rejection after organ transplantations. It is a 23-membered macrolide lactone and contains tricarbonyl and l-pipecolic acid moieties.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Evaluation of a Bio-solid phase microextraction-tandem mass spectrometry (SPME-MS/MS) method to determine four immunosuppressive drugs in whole human blood
  • High-performance liquid chromatographic (HPLC) analysis of tacrolimus tablet samples, prepared by freeze-drying
  • Development and validation of a reversed phase-high performance liquid chromatographic (RP-HPLC) for the determination of tacrolimus in its capsule formulations
  • Stability analysis of tacrolimus and simultaneous determination of its important impurities by reversed phase-ultra-high performance liquid chromatography (RP-UHPLC)

Biochem/physiol Actions

FK-506 is a potent immunosuppressant, neuroprotective and neuroregenerative, and in vitro T cell proliferation blocker. FK-506 disrupts calcineurin-mediated signal transduction in T lymphocytes and interacts with its FK-506-binding protein-12 (FKBP12).
FK-506 is a potent immunosuppressant; neuroprotective and neuroregenerative; in vitro T cell proliferation blocker.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1117 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversed-phase high performance liquid chromatographic (RP-HPLC) method for determination of tacrolimus in bulk and pharmaceutical formulation
Patel P, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 3, 220-222 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service