Skip to Content
Merck
All Photos(1)

Documents

84850

Sigma-Aldrich

Sebacoyl chloride

purum, ≥95.0% (GC)

Synonym(s):

Sebacyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO(CH2)8COCl
CAS Number:
Molecular Weight:
239.14
Beilstein:
1365665
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

purum

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.468 (lit.)
n20/D 1.468

bp

168 °C/12 mmHg (lit.)

mp

−5-−3 °C (lit.)

density

1.119 g/mL at 20 °C
1.121 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCCCCCC(Cl)=O

InChI

1S/C10H16Cl2O2/c11-9(13)7-5-3-1-2-4-6-8-10(12)14/h1-8H2

InChI key

WMPOZLHMGVKUEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sebacoyl chloride is a light yellow, pungent odored di-acryl chloride. It is soluble in hydrocarbons and ethers.It hydrolyzes in water, releasing hydrogen chloride.

Application

Sebacoyl chloride may be used along with hexamethylenediamine, during Schotten -Baumann reactions, to produce a polyamide (eg.Nylon 6,10).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Gong et al.
ACS biomaterials science & engineering, 6(11), 6331-6343 (2021-01-16)
Despite decades of research, spinal cord injury (SCI) still causes irreparable damage to the human body. Key challenges that hinder the regeneration and extension of neurons following SCI must be overcome, including the overexpressed glial scar formation and strong inflammatory
P L Madan et al.
Pharmaceutical research, 6(8), 714-718 (1989-08-01)
The effect of selected variables on the release characteristics of theophylline microcapsules prepared with 1,6-hexamethylene diamine and sebacoyl chloride via interfacial polycondensation has been investigated. The nature of the microcapsules and the release characteristics of theophylline from the microcapsules were
D Poncelet et al.
Journal of microencapsulation, 11(1), 31-40 (1994-01-01)
A microencapsulation technique is proposed involving the formation of a polyethyleneimine (PEI) membrane crosslinked by an acid dichloride. The membranes were formed at pH 8 in a non-polar solvent, conditions which are better suited for the encapsulation of biocatalysts or
Zhenhua Xu et al.
International journal of pharmaceutics, 426(1-2), 182-192 (2012-01-24)
In this study we describe a novel polymer, mPPS-FA, synthesized as a potential gene transfer vector. To complete mPPS-FA, folic acid was conjugated to a backbone (named mPPS) consisting of a copolymer of methyl PEG-2000, PEI-600, and sebacoyl chloride. (1)H
Marianna Sikorska et al.
BioFactors (Oxford, England), 18(1-4), 173-183 (2003-12-30)
We have derivatised alpha-tocopherol (vitamin E) to a water-soluble polyoxyethanyl-alpha - tocopheryl sebacate (PTS) and discovered that it formed a non-covalent complex with CoQ10 at a molar ratio of 2:1 (PTS-CoQ10). This complex was water-soluble and remained stable for extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service