40608
Schwarz Differential Agar
suitable for microbiology, NutriSelect® Plus
Synonym(s):
SDA, SDM, Schwarz Differential Medium
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 20 g/L
basic fuchsin, 0.47 g/L
dextrin, 0.11 g/L
dextrose, 10 g/L
malt extract, 3 g/L
peptic digest of animal tissue, 5 g/L
sodium sulfite, 2.92 g/L
yeast extract, 3 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
6.9±0.2 (25 °C)
application(s)
bioburden testing
food and beverages
microbiology
suitability
selective and differential for Candida spp.
selective and differential for Pichia spp.
selective and differential for Saccharomyces spp.
selective and differential for Zygosaccharomyces spp.
Related Categories
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.
Protocols
Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.
Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.
Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.
Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service