Skip to Content
Merck
All Photos(1)

Documents

34726BC

Millipore

Assurance® GDS DNA Swab Resuspension Buffer Tq PCR Reagent

For use with Assurance® GDS

Synonym(s):

Buffer for PCR Sample Prep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106306
NACRES:
NA.85

product name

GDS DNA Swab Resuspension Buffer Tq, BioControl, For use with GDS

Quality Level

shelf life

24 mo.

packaging

vial of 1.2 mL (sufficientfor 100 tests)

manufacturer/tradename

BioControl Systems 34726-100

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)
Cronobacter spp.
Salmonella spp.
enterovirulent E. coli (EEC) (Shiga Toxigenic)

General description

GDS DNA Swab Resuspension Buffer Tq is a requisite recommended for the labs to have onsite, in case of a contamination event. Improper handling and disposal of used Assurance® GDS amplification tubes may result in DNA amplicon contamination of laboratory work surfaces and equipment. As a preventive measure, it is recommended to perform periodic sampling for DNA contamination. GDS DNA Swab Resuspension Buffer Tq is transferred to the appropriate amplification tubes for all Assurance® GDS assays (except Listeria spp. and Listeria monocytogenes).

Application

GDS DNA Swab Resuspension Buffer Tq is used during environmental sampling for DNA contamination.

Components

One vial, 1.2 mL, sufficient for 100 tests

Compatibility

BioControl GDS

Testing Method

PCR

Other Notes

Resuspension Buffer Tq from GDS kits CANNOT be substituted for DNA Swab Resuspension Buffer Tq.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service