SCT084
BioTracker™ MemBright 560 Live Cell Dye
Synonym(s):
Live cell imaging probe, Live cell membrane probe, orange/red
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
mol wt
observed mol wt 1370.3 kDa
Quality Level
packaging
vial of 50 μL
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell based assay: suitable
flow cytometry: suitable
detection method
fluorometric
shipped in
dry ice
storage temp.
−20°C
General description
The BioTracker™ Membright-560 live cell dye is a “turn-on” fluorescence probe emitting in the orange to red spectrum for imaging of plasma membranes of live cells.
Probes specific for the plasma membrane are essential for demarcating cell boundaries. Membright dyes use a novel fluorogenic mechanism, operating in a self-quenched nanoparticle state. On contact and propagation within the cell membrane, fluorescence is turned on, producing a bright signal with low to no background. The Membright live cell membrane probes are compatible with two-photon imaging, superresolution microscopy, and can be used in live or fixed cells. Their robust photostability and lack of cytotoxicity make Membright probes optimal for protracted live cell imaging. The superior characteristics of these plasma membrane probes permit use in a diverse spectrum of fluorescence detection applications.
Spectral Properties
Fluorescence images obtained by λex = 560 nm and emission range of 567-750 nm.
Reference
Collot M et al. MemBright: A family of fluorescent membrane probes for advanced cellular imaging and neuroscience. Cell Chem. Biol. 2019, 26:600-614
Probes specific for the plasma membrane are essential for demarcating cell boundaries. Membright dyes use a novel fluorogenic mechanism, operating in a self-quenched nanoparticle state. On contact and propagation within the cell membrane, fluorescence is turned on, producing a bright signal with low to no background. The Membright live cell membrane probes are compatible with two-photon imaging, superresolution microscopy, and can be used in live or fixed cells. Their robust photostability and lack of cytotoxicity make Membright probes optimal for protracted live cell imaging. The superior characteristics of these plasma membrane probes permit use in a diverse spectrum of fluorescence detection applications.
Spectral Properties
Fluorescence images obtained by λex = 560 nm and emission range of 567-750 nm.
Reference
Collot M et al. MemBright: A family of fluorescent membrane probes for advanced cellular imaging and neuroscience. Cell Chem. Biol. 2019, 26:600-614
Application
Purity: > 98% confirmed by HPLC, HNMR, LC-MS and elemental analysis.
Features and Benefits
Red/orange fluorescent probe for efficient, selective labeling of the plasma membrane in live cells. Suitable for superresolution microscopy.
Physical form
Magenta solution
Storage and Stability
Store long term at -20°C, protected from light.
Other Notes
Live cell fluorescent imaging, flow cytometry
Legal Information
BioTracker is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service