Skip to Content
Merck
All Photos(4)

Key Documents

MABF3125

Sigma-Aldrich

Anti-Glucuronoxylomannan/GXM Antibody, clone 3E5

Synonym(s):

GXM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3E5, monoclonal

mol wt

observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

fungi

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
neutralization: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Specificity

Clone 3E5 is a mouse monoclonal antibody that detects Glucuronoxylomannan /GXM in Cryptococcus neoformans infected mouse cells.

Immunogen

Glucuronoxylomannan/GXM from Cryptococcus neoformans conjugated to tetanus toxoid.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Cryptococcus neoformans infected Mouse lung tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected glucuronoxylomannan/GXM in in Cryptococcus neoformans infected Mouse lung tissue sections.

Tested Applicatins

Neutralizing: A representative lot neutralized Gonoxylomannan/GXM and prevented toxicity in mice. (Lendvai, N., et al. (1999). J Infect Dis.180(3):791-801).

Immunohistochemistry Applications: A representative lot detected Glucuronoxylomannan/GXM in Immunohistochemistry applications (Feldmesser, M., et al. (1997). J Immunol. 158(2):790-9; Lendvai, N., et al. (1999). J Infect Dis. 180(3):791-801).

Fluorescence Activated Cell Sorting (FACS): A representative lot was used for sorting J774 and CHO cells labeled with this antibody in Fluorescence Activated Cell Sorting applications (Saylor Hawk, C., et al. (2019). J Immunol. 202(9):2782-2794).

ELISA Analysis: A representative lot detected Glucuronoxylomannan/GXM in ELISA applications (Janda, A., et al. (2015). J Biol Chem. 290(19):12090-100; Saylor Hawk, C., et al. (2019). J Immunol. 202(9):2782-2794).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Glucuronoxylomannan (GXM), a polysaccharide, is the principal constituent of the Cryptococcus neoformans capsule that modulates the inflammatory response of human monocytes in vitro and exerts many immunoregulatory effects. Its basic structure is a tri-mannose repeat with a glucuronic acid residue in the first mannose. Its structure is shown to be further modified in individual strains by the addition of xylose substitutions on the mannan backbone. Six triads known as M1-M2 are shown to be present in various proportions in GXM from the various serotypes and M2 is reported to be the most common triad in serotype A GXM. Soluble GXM serves as an important virulence factor and contributes to the pathogenesis of infection. High levels of GXM are found in serum of patients with cryptococcosis. Clone 3E5 is a class II monoclonal Abs that is reported to possess hydrolytic activity against a 12 amino acid P1 peptide (SPNQHTPPWMLK) at positions 1, 2, and 10. (Ref.: Bowen, A., et al. (2017). J. Biol. Chem. 292(2); 417-434; Tissi, L., et al. (2004). Infect. Immun. 72(11); 6367-6372; Lendvai, N., and Casadevall, A. (1999). J. Infect. Dis. 180(3); 791-801).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service