Skip to Content
Merck
All Photos(1)

Key Documents

FR123696087

Sigma-Aldrich

Estapor® White Carboxylated microspheres

K1-080 White COOH PS 10%, White carboxylated microspheres 800nm, Latex Turbidimetric Immunoassay - For ordering, click "Request more information"

Synonym(s):

Carboxylic Beads, Carboxylic Particles, Plain Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102026
NACRES:
NA.24

Quality Level

manufacturer/tradename

Estapor®

packaging

bottle of 10ml-10L

concentration

10% solids

technique(s)

latex turbidmetry immunoassay (LTIA): suitable

dilution

(Latex Agglutination Test (LAT))

color

white

matrix

polymeric Poly-styrene-co-acrylate

bead diameter

0.76-0.94 μm±<5%

capacity

1-10 μeq/g surface charge density

functional group

carboxylic acid (COOH)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Estapor® carboxylated white microspheres available in different polymer matrixes, sizes and carboxyl content offer are vesatlie options for the development of a wide range Latex Turbidimetry Immunoassay (LTIA) diagnostic tests. The carboxylated surface of the microsphere is designed to be coated with the antibody or antigen of choice via a standard 2 steps EDC/NHS conjugation protocol. The robustness of the covalent bond of this conjugation process ensures long term stability and results reliability. This is a key feature of these microspheres and has attracted many diagnostic test developers whom have successfully utilized these microspheres for several years in a wide variety of LTIAs.

Application

Main applications of the standard microspheres include nephelemetric assays, immune-turbidimetric assays, latex agglutination tests, solid phase immunoassays, microspheres capture ELISA, calibration

Legal Information

Estapor is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service