Skip to Content
Merck
All Photos(8)

Key Documents

EQ70XXPKIT

Milli-Q

Milli-Q® EQ 7008/16 Ultrapure Water Purification Kit

new
greener alternative

For Milli-Q® EQ 7008/16 systems to produce RO (Type 3) and ultrapure (Type 1) water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200

Quality Level

packaging

units of 1

manufacturer/tradename

Milli-Q®

storage condition

room temperature

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

cartridge H

39.5 cm (15.5 in.)

input

feed water nature tap water (within specifications of the water purification system)

compatibility

for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016

greener alternative category

General description

This water purification kit includes:
  • IPAK Gard® 3/5 pretreatment cartridge is composed of a pleated filter for the removal of particulates, minerals and colloids, and a carbon block for the removal of free chlorine from tap feed water. This protects the reverse osmosis (RO) membrane and improves system performance.
  • IPAK Meta® and IPAK Quanta® polishing cartridges for purified water, designed to work as a pair to produce ultrapure (Type 1) water for sensitive laboratory applications. The cartridges contain a combination of Jetpore® mixed-bed ion-exchange resin and innovative IQnano ion-exchange media to remove trace ions from purified feed water, as well as a high-grade synthetic activated carbon, which targets trace organic contaminants.
  • The Tank Vent Filter protects stored RO (Type 3) water from airborne contaminants. It includes activated carbon, soda lime and a membrane filter to prevent impure air from entering the tank.

We continue to demonstrate our commitment to a sustainable future by exploring ways to improve existing products in our portfolio. We have reviewed our purification cartridges to identify key opportunities to minimize impact without compromising on the product performance. This product has benefited from an optimization of the resin mix, the packaging and the end-of-life management. Therefore, it is identified as a greener alternative product in the category reengineered. View the DfS scorecard for the new Milli-Q® EQ 7008/16 purification kit.

Application

  • For Milli-Q® EQ 7008/16 ultrapure water purification systems
  • Optimally protects RO membranes (IPAK Gard®), removes trace ions and organics (Milli-Q® polishing cartridges), and protect stored water from airborne contaminants (Tank Vent Filter)

Features and Benefits

  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with one Drain Cap per cartridge, to remove excess water and reduce weight of exhausted cartridges at disposal.
  • 63% of the parts suppliers for IPAK Gard® cartridge and 78% of the parts suppliers for IPAK Quanta® and IPAK Meta® cartridges included in this kit are ISO 14001 environmental management system-certified.

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service