Skip to Content
Merck
All Photos(2)

Key Documents

ABE40

Sigma-Aldrich

Anti-PABPC1/C3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Polyadenylate-binding protein 1, PABP-1, Poly(A)-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
mouse, human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

canine (based on 100% sequence homology), bovine (based on 100% sequence homology), chicken (based on 100% sequence homology), rat (based on 100% sequence homology), Xenopus (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PABPC1(26986)

General description

Poly(A)-binding proteins, cytoplasmic 1 (PABPC1), and cytoplasmic 3 (PABPC3) are RNA-binding proteins with important roles in post-translational control and metabolism of mRNA. Although both proteins bind to the poly(A) tail of mRNA, PABPC3 binds with a lower affinity than PABPC1. Studies have associated these proteins with tumorigenesis. Decreased expression of PABPC1 in esophageal cancer seems to associate with tumor development and poor prognosis after surgery. PABPC1 can shuttle through the nuclear membrane and is thought to associate with newly synthesized poly(A) transcripts. PABPC1 has also been linked to microRNA mediated silencing when in contact with GW182. PABPC3 is thought to be specific to human testis and controls RNA stability.

Immunogen

KLH-conjugated linear peptide corresponding to human PABPC1/C3. The immunogen sequence used is 100% identical to both PABPC1 and PABPC3.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected PABPC1/C3 in NIH/3T3 and HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-PABPC1/C3 (Rabbit Polyclonal Antibody) validated in ICC, WB to detect PABPC1/C3 also known as Polyadenylate-binding protein 1, PABP-1.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PABPC1/C3 on 10 µg of HeLa cell lysate.

Target description

~71 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulino Barragán-Iglesias et al.
Nature communications, 9(1), 10-10 (2018-01-04)
Nociceptors rely on cap-dependent translation to rapidly induce protein synthesis in response to pro-inflammatory signals. Comparatively little is known regarding the role of the regulatory factors bound to the 3' end of mRNA in nociceptor sensitization. Poly(A)-binding protein (PABP) stimulates translation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service