Skip to Content
Merck
All Photos(1)

Key Documents

8.18704

Sigma-Aldrich

Cetyl alcohol

for synthesis

Synonym(s):

Cetyl alcohol, 1-Hexadecanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
MDL number:
UNSPSC Code:
12352104
EC Index Number:
253-149-0
NACRES:
NA.22
grade:
for synthesis
bp:
305-330 °C/1013 hPa
vapor pressure:
<1 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

<1 hPa ( 20 °C)

Assay

≥95% (GC)

form

solid

autoignition temp.

235 °C (DIN 51794)

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

bp

305-330 °C/1013 hPa

mp

46-52 °C

transition temp

flash point 155 °C (DIN 51758)

solubility

<0.0001 g/L

density

0.82 g/cm3 at 50 °C

bulk density

410‑450 kg/m3

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCCCCCCC

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Melting range (lower value): ≥ 47 °C
Melting range (upper value): ≤ 52 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service