Skip to Content
Merck
All Photos(1)

Key Documents

1.13760

Millipore

Sodium dodecyl sulfate

LAB

Synonym(s):

Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Dodecyl sodium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NaO4S
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-788-1
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Dodecyl sulfate sodium salt, LAB

Quality Level

potency

1427 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

6-9 (20 °C, 10 g/L in H2O)

mp

204-207 °C

solubility

>130 g/L

density

1.1 g/cm3 at 20 °C

bulk density

490‑560 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Analysis Note

Assay (two-phase titration): ≥ 99.0 %
Identity (IR-spectrum): passes test
Absorbance A (220-350 nm; 3 %; 1 cm; water): ≤ 0.1
Chloride (Cl): ≤ 0.01 %
Phosphate (PO₄): ≤ 0.0001 %
Heavy metals (as Pb): ≤ 0.0005 %
Water (according to Karl Fischer): ≤ 1

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service