Skip to Content
Merck
All Photos(1)

Documents

T-008

Supelco

(±)-11-nor-9-Carboxy-Δ9-THC-D3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25O4D3
CAS Number:
Molecular Weight:
347.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

OC1=C2C3C(CCC(C(O)=O)=C3)C(C)(C)OC2=CC(CCCCC([2H])([2H])[2H])=C1

InChI

1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h10-12,15-16,22H,4-9H2,1-3H3,(H,23,24)/i1D3

InChI key

YOVRGSHRZRJTLZ-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

11-nor-9-Carboxy-Δ9-THC, also known as THC acid, is the main secondary metabolite of Δ9-THC found in urine or blood of cannabis users. This internal standard is applicable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug testing and isotope dilution methods.

Application


  • Integration of an online application to optimize multi-disciplinary meetings: This study implemented an online application designed to optimize the scheduling and management of multi-disciplinary meetings in healthcare settings, showcasing its applications in streamlining communication and decision-making processes (Magee et al., 2024).

  • Determining a musculoskeletal system′s pre-stretched state: Research demonstrated the use of continuum-mechanical forward modeling and joint range optimization in determining the pre-stretched state of musculoskeletal systems, offering insights into biomechanical applications and potential improvements in clinical assessments (Avci and Röhrle, 2024).

  • From leaf to multiscale models of photosynthesis: This review discusses the applications of multiscale modeling from individual leaves to entire crops in photosynthesis research, highlighting its challenges and opportunities in enhancing crop yields and photosynthetic efficiency (Stirbet et al., 2024).

  • Syngas production from CO₂ and H₂O via solid-oxide electrolyzer cells: The article covers the comprehensive application of solid-oxide electrolyzer cells for syngas production, focusing on the fundamentals, materials, degradation, and operating conditions. It reflects on the broader applicability in sustainable energy solutions (Hou et al., 2024).

  • Comparison of mechanochemical methods in the synthesis of binaphthol-benzoquinone based cocrystals: This study compares various mechanochemical methods for synthesizing cocrystals, providing valuable insights into pharmaceutical applications and material engineering (Nagapudi and Nagapudi, 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service