Skip to Content
Merck
All Photos(1)

Key Documents

800605O

Avanti

18:1 Ethylene Glycol

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl) ethylene glycol; DEG(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H70O4
CAS Number:
Molecular Weight:
590.96
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

1-2-dioleoyl ethylene glycol, neat oil

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 10 mg (800605O-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(CCOC(=O)CCCCCCC\C=C\CCCCCCCC)C(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C38H70O4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37(39)41-35-36-42-38(40)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-36H2,1-2H3/b19-17+,20-18+

InChI key

NKSOSPOXQKNIKJ-XPWSMXQVSA-N

General description

18:1 Ethylene glycol (1-2-dioleoyl ethylene glycol), a nonpolar lipid, contains two oleic acid esterified to ethylene glycol. Ethylene glycol is the major polyol for oleic acid esters synthesis.

Biochem/physiol Actions

18:1 Ethylene Glycol or dioleoyl ethylene glycol (DOEG) is protein kinase C inhibitor.

Packaging

5 mL Clear Glass Sealed Ampule (800605O-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shawn Hopewell et al.
Metabolism: clinical and experimental, 57(12), 1677-1684 (2008-11-18)
Phosphatidylinositol (PI) has been shown to stimulate reverse cholesterol transport in animal models and to increase plasma apolipoprotein (apo) A-I levels and high-density lipoprotein cholesterol in human subjects. The objective of this study was to determine the molecular mechanism through
Asma Chaari et al.
Journal of oleo science, 66(5), 455-461 (2017-04-07)
The production of fatty acid esters from stearic, oleic, and palmitic acids and polyols (ethylene glycol and glycerol) was investigated in this work. A series of montmorillonite-based clays catalysts (KSF, KSF/0, KP10, and K10), having different physicochemical properties, were used
Michal Holčapek et al.
Journal of chromatography. A, 1377, 85-91 (2014-12-30)
A novel normal-phase (NP) ultrahigh-performance liquid chromatography-mass spectrometry (UHPLC/MS) method is developed for a separation and quantitation of nonpolar lipid classes occurring in human plasma, erythrocytes and plasma lipoprotein fractions. The baseline class separation of cholesteryl esters (CE), cholesterol, triacylglycerols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service