Skip to Content
Merck
All Photos(1)

Key Documents

640008P

Avanti

Girard Reagent-d5

Avanti Research - A Croda Brand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5D5BrN3O
Molecular Weight:
237.11
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (640008P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(NN)C[N+]1=C([2H])C([2H])=C([2H])C([2H])=C1[2H].[Br-]

InChI

1S/C7H9N3O.BrH/c8-9-7(11)6-10-4-2-1-3-5-10;/h1-5H,6,8H2;1H/i1D,2D,3D,4D,5D;

InChI key

URTAUWVMJKDFIX-GWVWGMRQSA-N

General description

Girard Reagents are N-substituted glycine hydrazides. They are classified as Girard-T (trimethylacetylhydrazide ammonium chloride), Girard-P (pyridinioacetohydrazide chloride) and Girard-D (N,N-dimethylglycine hydrazide hydrochloride). Girard Reagent-d5 is an isotope containing labeling reagent.

Biochem/physiol Actions

Girard reagents generate water-soluble hydrazones of carbonyl compounds aiding their separation. These reagents are sensitive and selective. d5-Girard reagent P (d5-GP) is useful in the labeling of steroid hormones for mass spectroscopy analysis enabling accurate cum sensitive steroid hormone quantification.

Packaging

5 mL Amber Glass Screw Cap Vial (640008P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transition metal complexes with Girard reagents and their hydrazones
Vojinovic-Jevsic LS, et al.
J. Serb. Chem. Soc., 77(9), 1129-1155 (2012)
Ning Guo et al.
Analytica chimica acta, 905, 106-114 (2016-01-13)
Steroid hormones play important roles in mammal at very low concentrations and are associated with numerous endocrinology and oncology diseases. Therefore, quantitative analysis of steroid hormones can provide crucial information for uncovering underlying mechanisms of steroid hormones related diseases. In
Highly automated nano-LC/MS-based approach for thousand cell-scale quantification of side chain-hydroxylated oxysterols
Roberg-Larsen H, et al.
Journal of Lipid Research, 55(7), 1531-1536 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service