Z743894
ART® SoftFit L Reload System
Pure 1000L, size 1000 μL, reload insert, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
sterility
sterile
feature
Pure 1000L
reload insert
packaging
pkg of 4X768 ea
manufacturer/tradename
Molecular BioProducts 3781-RI
technique(s)
PCR: suitable
size
1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
The ART™ SoftFit-L Reload System is a complete product offering of pipet tips compatible with LTS™ pipetters. This unique system provides preloaded pipet tips in color-coded inserts by volume size with the option of an easy-to-use ′push down′ reload pack. Choose sterile or non-sterile tips, in five volume sizes. Pipette samples with the accuracy and precision you expect from your pipette, every time.
Features and Benefits
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service