Z743890
ART® SoftFit L Reload System
HLT 300L, size 300 μL, reload insert, non-sterile
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
sterility
non-sterile
feature
HLT 300L
reload insert
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3732-RI
technique(s)
PCR: suitable
size
300 μL
Looking for similar products? Visit Product Comparison Guide
General description
The ART™ SoftFit-L Reload System is a complete product offering of pipet tips compatible with LTS™ pipetters. This unique system provides preloaded pipet tips in color-coded inserts by volume size with the option of an easy-to-use ′push down′ reload pack. Choose sterile or non-sterile tips, in five volume sizes. Pipette samples with the accuracy and precision you expect from your pipette, every time.
Features and Benefits
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service