Z677337
Dermatril™ LR nitrile gloves
size L
Synonym(s):
disposable glove
All Photos(1)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
blue gloves
blue nitrile
size
L
sterility
non-sterile
feature
CE compliant
latex free
powder-free
packaging
pack of 50 ea (boxes of 50)
manufacturer/tradename
KCL 742 - 09
KCL 742
L × thickness
270 mm × 0.11 mm
General description
Dermatril LR is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril LR disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril LR disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
All Dermatril LR disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
- Cleanroom class 100
- Anti-static
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- Extended cuff
- Roughened fingertips for better grip
- Rolled cuff edge for easier donning
- CE compliant
- Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001, US-FED-ST 209E
Legal Information
Dermatril is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)