Key Documents
TEM-FF400CU
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 400 mesh, box of 25
Synonym(s):
FORMVAR FILM 400# COPPER 25/PK
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
400 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 400 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)