DWKBWP0500AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap
round amber polypropylene bottle, capacity 500 mL
Synonym(s):
BWP0500AP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
round amber polypropylene bottle
wide-neck
packaging
pkg of 10 ea
manufacturer/tradename
Azlon BWP0500AP
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Application
Ideal for a wide range of light sensitive liquid or powder applications
Features and Benefits
Autoclavable at 121°C and suitable for food contact applications
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service